Product Details

SNP ID
rs79050107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8693527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTCGGAGGGTGGGAGGAATGA[C/G]AGGACTAGGCCTAAGACCCTGGTGC
Phenotype
MIM: 603637 MIM: 611333 MIM: 140750
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RPL27A PubMed Links

Gene Details

Gene
RPL27A
Gene Name
ribosomal protein L27a
There are no transcripts associated with this gene.

Gene
SNORA3A
Gene Name
small nucleolar RNA, H/ACA box 3A
There are no transcripts associated with this gene.

Gene
SNORA3B
Gene Name
small nucleolar RNA, H/ACA box 3B
There are no transcripts associated with this gene.

Gene
ST5
Gene Name
suppression of tumorigenicity 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005418.3 4377 UTR 3 NP_005409.3
NM_139157.2 4377 UTR 3 NP_631896.1
NM_213618.1 4377 UTR 3 NP_998783.1
XM_005253077.1 4377 UTR 3 XP_005253134.1
XM_005253083.2 4377 UTR 3 XP_005253140.1
XM_011520309.1 4377 UTR 3 XP_011518611.1
XM_011520310.1 4377 UTR 3 XP_011518612.1
XM_011520311.2 4377 UTR 3 XP_011518613.1
XM_011520312.1 4377 UTR 3 XP_011518614.1
XM_011520313.1 4377 UTR 3 XP_011518615.1
XM_011520314.1 4377 UTR 3 XP_011518616.1
XM_011520315.1 4377 UTR 3 XP_011518617.1
XM_011520316.1 4377 UTR 3 XP_011518618.1
XM_011520317.1 4377 UTR 3 XP_011518619.1
XM_011520318.1 4377 UTR 3 XP_011518620.1
XM_011520319.2 4377 UTR 3 XP_011518621.1
XM_011520320.1 4377 Intron XP_011518622.1
XM_011520321.2 4377 Intron XP_011518623.1
XM_011520322.1 4377 Intron XP_011518624.1
XM_011520323.2 4377 UTR 3 XP_011518625.2
XM_011520324.2 4377 UTR 3 XP_011518626.2
XM_011520325.1 4377 UTR 3 XP_011518627.1
XM_011520326.1 4377 UTR 3 XP_011518628.1
XM_011520327.1 4377 UTR 3 XP_011518629.1
XM_011520328.1 4377 UTR 3 XP_011518630.1
XM_011520329.1 4377 UTR 3 XP_011518631.1
XM_017018182.1 4377 UTR 3 XP_016873671.1
XM_017018183.1 4377 UTR 3 XP_016873672.1
XM_017018184.1 4377 UTR 3 XP_016873673.1
XM_017018185.1 4377 UTR 3 XP_016873674.1
XM_017018186.1 4377 UTR 3 XP_016873675.1
XM_017018187.1 4377 UTR 3 XP_016873676.1

View Full Product Details