Product Details

SNP ID
rs79712146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75003603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATTGGAATGCAGGTTGGCTGGT[G/A]CCAGAACCTGGGCTCTTAACCAGTA
Phenotype
MIM: 612060
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ZNRF1 PubMed Links

Gene Details

Gene
ZNRF1
Gene Name
zinc and ring finger 1, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032268.4 Intron NP_115644.1
XM_011523392.1 Intron XP_011521694.1
XM_017023793.1 Intron XP_016879282.1
XM_017023794.1 Intron XP_016879283.1

View Full Product Details