Product Details

SNP ID
rs78069344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48912713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTTCCTTCTGCATGGTGAGGAT[A/G]TAGAACATGTGTACCAGATTATGGA
Phenotype
MIM: 611362
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105371814 PubMed Links

Gene Details

Gene
LOC105371814
Gene Name
uncharacterized LOC105371814
There are no transcripts associated with this gene.

Gene
UBE2Z
Gene Name
ubiquitin conjugating enzyme E2 Z
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023079.4 Intron NP_075567.2

View Full Product Details