Product Details

SNP ID
rs75698710
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:67852119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTTTAATTTAATGTCGTTCTCT[C/A]TTTAAACACTGTAGTCTATGTTCAA
Phenotype
MIM: 601819
Polymorphism
C/A, Transversion Substitution
Allele Nomenclature
Literature Links
BPTF PubMed Links

Gene Details

Gene
BPTF
Gene Name
bromodomain PHD finger transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004459.6 Intron NP_004450.3
NM_182641.3 Intron NP_872579.2
XM_005257150.3 Intron XP_005257207.1
XM_005257151.3 Intron XP_005257208.1
XM_005257152.2 Intron XP_005257209.1
XM_005257153.3 Intron XP_005257210.1
XM_005257154.3 Intron XP_005257211.1
XM_005257155.3 Intron XP_005257212.1
XM_005257156.3 Intron XP_005257213.1
XM_005257157.3 Intron XP_005257214.1
XM_005257158.3 Intron XP_005257215.1
XM_005257159.2 Intron XP_005257216.1
XM_005257160.2 Intron XP_005257217.1
XM_005257161.3 Intron XP_005257218.1
XM_011524520.2 Intron XP_011522822.1
XM_011524521.2 Intron XP_011522823.1
XM_011524522.2 Intron XP_011522824.1
XM_011524523.2 Intron XP_011522825.1
XM_011524524.2 Intron XP_011522826.1
XM_011524525.2 Intron XP_011522827.1
XM_011524526.2 Intron XP_011522828.1
XM_017024353.1 Intron XP_016879842.1
XM_017024354.1 Intron XP_016879843.1

View Full Product Details