Product Details

SNP ID
rs79756729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:72647277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGGAGTTGGGAGGCAGATAGAA[C/G]GTCCCGAAGTGGAGATTTCCCTGAT
Phenotype
MIM: 616508
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00511 PubMed Links

Gene Details

Gene
LINC00511
Gene Name
long intergenic non-protein coding RNA 511
There are no transcripts associated with this gene.

Gene
SLC39A11
Gene Name
solute carrier family 39 member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159770.1 1427 UTR 3 NP_001153242.1
NM_139177.3 1427 UTR 3 NP_631916.2
XM_005257134.3 1427 UTR 3 XP_005257191.1
XM_006721754.3 1427 UTR 3 XP_006721817.1
XM_006721755.2 1427 UTR 3 XP_006721818.1
XM_006721756.2 1427 Intron XP_006721819.1
XM_006721757.2 1427 UTR 3 XP_006721820.1
XM_006721758.3 1427 Intron XP_006721821.1
XM_006721759.1 1427 Intron XP_006721822.1
XM_011524493.1 1427 UTR 3 XP_011522795.1
XM_011524494.2 1427 UTR 3 XP_011522796.1
XM_011524495.2 1427 Intron XP_011522797.1
XM_011524496.2 1427 Intron XP_011522798.1
XM_011524498.2 1427 Intron XP_011522800.1
XM_017024330.1 1427 UTR 3 XP_016879819.1
XM_017024331.1 1427 UTR 3 XP_016879820.1
XM_017024332.1 1427 UTR 3 XP_016879821.1
XM_017024333.1 1427 UTR 3 XP_016879822.1
XM_017024334.1 1427 UTR 3 XP_016879823.1
XM_017024335.1 1427 UTR 3 XP_016879824.1
XM_017024336.1 1427 UTR 3 XP_016879825.1
XM_017024337.1 1427 UTR 3 XP_016879826.1
XM_017024338.1 1427 Intron XP_016879827.1
XM_017024339.1 1427 Intron XP_016879828.1
XM_017024340.1 1427 Intron XP_016879829.1
XM_017024341.1 1427 Intron XP_016879830.1
XM_017024342.1 1427 Intron XP_016879831.1

View Full Product Details