Product Details

SNP ID
rs78428884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:2573995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATGGAAATCATAATGTAGATAG[G/A]GTTTTTGTTCTGCAGAGAAGCAAAA
Phenotype
MIM: 607272
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
METTL4 PubMed Links

Gene Details

Gene
METTL4
Gene Name
methyltransferase like 4
There are no transcripts associated with this gene.

Gene
NDC80
Gene Name
NDC80, kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006101.2 Intron NP_006092.1

View Full Product Details