Product Details

SNP ID
rs77138444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43952285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACACAACCATGGTATTTTGTAGC[A/G]TTGTTTCCGCATCCTGACTTCTCTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF221 PubMed Links

Gene Details

Gene
ZNF221
Gene Name
zinc finger protein 221
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297588.1 Intron NP_001284517.1
NM_001297589.1 Intron NP_001284518.1
NM_013359.2 Intron NP_037491.2
XM_017027232.1 Intron XP_016882721.1
XM_017027233.1 Intron XP_016882722.1

View Full Product Details