Product Details

SNP ID
rs77692880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:10229614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAACTCTACAGACAGGAGAGCTT[A/G]CTGGGGAAAGATTTTGAAAGTCACA
Phenotype
MIM: 600322
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNAP25 PubMed Links

Gene Details

Gene
SNAP25
Gene Name
synaptosome associated protein 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322902.1 Intron NP_001309831.1
NM_001322903.1 Intron NP_001309832.1
NM_001322904.1 Intron NP_001309833.1
NM_001322905.1 Intron NP_001309834.1
NM_001322906.1 Intron NP_001309835.1
NM_001322907.1 Intron NP_001309836.1
NM_001322908.1 Intron NP_001309837.1
NM_001322909.1 Intron NP_001309838.1
NM_001322910.1 Intron NP_001309839.1
NM_003081.4 Intron NP_003072.2
NM_130811.3 Intron NP_570824.1
XM_005260808.4 Intron XP_005260865.1
XM_017028021.1 Intron XP_016883510.1
XM_017028022.1 Intron XP_016883511.1
XM_017028023.1 Intron XP_016883512.1

View Full Product Details