Product Details

SNP ID
rs77391194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:4732383 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACATGGAGCTGCCACTGGAGTT[C/G]AGGATTATTTAGCAGTAAATCTTTT
Phenotype
MIM: 604263
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRND PubMed Links

Gene Details

Gene
PRND
Gene Name
prion protein 2 (dublet)
There are no transcripts associated with this gene.

Gene
PRNT
Gene Name
prion protein (testis specific)
There are no transcripts associated with this gene.

View Full Product Details