Product Details

SNP ID
rs76957251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:37209771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCACAAACATTGCAGCATTTAA[C/T]TTCCCCAAGTACCCTATGAAGAAGA
Phenotype
MIM: 602259
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSCR9 PubMed Links

Gene Details

Gene
DSCR9
Gene Name
Down syndrome critical region 9 (non-protein coding)
There are no transcripts associated with this gene.

Gene
TTC3
Gene Name
tetratricopeptide repeat domain 3
There are no transcripts associated with this gene.

View Full Product Details