Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001009554.3 | 5195 | UTR 3 | NP_001009554.1 | ||
NM_001301647.1 | 5195 | UTR 3 | NP_001288576.1 | ||
NM_001301648.1 | 5195 | UTR 3 | NP_001288577.1 | ||
NM_021647.7 | 5195 | UTR 3 | NP_067679.6 | ||
XM_005263366.3 | 5195 | UTR 3 | XP_005263423.1 | ||
XM_005263368.3 | 5195 | UTR 3 | XP_005263425.1 | ||
XM_017008864.1 | 5195 | UTR 3 | XP_016864353.1 | ||
XM_017008865.1 | 5195 | UTR 3 | XP_016864354.1 | ||
XM_017008866.1 | 5195 | UTR 3 | XP_016864355.1 | ||
XM_017008867.1 | 5195 | UTR 3 | XP_016864356.1 | ||
XM_017008868.1 | 5195 | UTR 3 | XP_016864357.1 | ||
XM_017008869.1 | 5195 | UTR 3 | XP_016864358.1 | ||
XM_017008870.1 | 5195 | UTR 3 | XP_016864359.1 | ||
XM_017008871.1 | 5195 | UTR 3 | XP_016864360.1 |