Product Details

SNP ID
rs76468558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27429887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGTATTAGAATTCTACTGTTAT[C/T]TCCTACTGTAGTCTGGCTGGGAATT
Phenotype
MIM: 606010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRBP1 PubMed Links

Gene Details

Gene
NRBP1
Gene Name
nuclear receptor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321357.1 Intron NP_001308286.1
NM_001321358.1 Intron NP_001308287.1
NM_001321359.1 Intron NP_001308288.1
NM_001321361.1 Intron NP_001308290.1
NM_001321362.1 Intron NP_001308291.1
NM_001321363.1 Intron NP_001308292.1
NM_013392.3 Intron NP_037524.1

View Full Product Details