Product Details

SNP ID
rs79945546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:14114144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAAGCCTGGCACTCTGCACTAAC[G/A]TCCAAAGTCCTAGGCGAAACCCCGC
Phenotype
MIM: 611077
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CHCHD4 PubMed Links

Gene Details

Gene
CHCHD4
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098502.1 Intron NP_001091972.1
NM_144636.2 Intron NP_653237.1

View Full Product Details