Product Details

SNP ID
rs75917388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:54124790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAACCTTATTCATTGAAAGATTG[C/G]TTGCTCAACTCTGTTGTTTGGGAAG
Phenotype
MIM: 606399
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CACNA2D3 PubMed Links

Gene Details

Gene
CACNA2D3
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018398.2 Intron NP_060868.2
XM_005265318.2 Intron XP_005265375.1
XM_011533946.2 Intron XP_011532248.1
XM_011533947.2 Intron XP_011532249.1
XM_011533948.2 Intron XP_011532250.1
XM_011533949.2 Intron XP_011532251.1
XM_011533950.2 Intron XP_011532252.1
XM_011533951.2 Intron XP_011532253.1
XM_011533952.2 Intron XP_011532254.1
XM_011533953.2 Intron XP_011532255.1
XM_011533954.2 Intron XP_011532256.1
XM_011533955.1 Intron XP_011532257.1
XM_017006850.1 Intron XP_016862339.1
XM_017006851.1 Intron XP_016862340.1
XM_017006852.1 Intron XP_016862341.1
XM_017006853.1 Intron XP_016862342.1
XM_017006854.1 Intron XP_016862343.1

View Full Product Details