Product Details
- SNP ID
-
rs75710828
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70029367 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATTTTTATAGTTAGTCCTATACA[A/G]GAAGTTTCCATTTAAATGAGTGAAG
- Phenotype
-
MIM: 142702
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HTN3
PubMed Links
Gene Details
- Gene
- HTN3
- Gene Name
- histatin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000200.2 |
|
Intron |
|
|
NP_000191.1 |
View Full Product Details