Product Details

SNP ID
rs77978711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:28822350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGACTGTTAAAGCCAAAATCCAG[A/G]AGGATGCCGAAACCCACTCCGGCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTUS2 PubMed Links

Gene Details

Gene
MTUS2
Gene Name
microtubule associated tumor suppressor candidate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033602.2 Intron NP_001028774.2
NM_015233.5 Intron NP_056048.1
XM_011535019.2 Intron XP_011533321.1
XM_011535020.2 Intron XP_011533322.1
XM_011535021.2 Intron XP_011533323.1
XM_011535022.2 Intron XP_011533324.1
XM_017020500.1 Intron XP_016875989.1
XM_017020501.1 Intron XP_016875990.1
XM_017020502.1 Intron XP_016875991.1

View Full Product Details