Product Details

SNP ID
rs76721111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63124654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCATAATAATTTAAAAAAAAAA[T/A]CCCTGAAAGATCCAAATTGAATAAC
Phenotype
MIM: 151430
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
BCL2 PubMed Links

Gene Details

Gene
BCL2
Gene Name
BCL2, apoptosis regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000633.2 5184 UTR 3 NP_000624.2
NM_000657.2 5184 Intron NP_000648.2
XM_011526135.2 5184 Intron XP_011524437.1
XM_017025917.1 5184 Intron XP_016881406.1

View Full Product Details