Product Details

SNP ID
rs75999880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149969318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGGGGATTGTAAGATGTGGCTT[G/T]AAGAATTTTTGATGTACTGGGAACA
Phenotype
MIM: 605697
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ULBP1 PubMed Links

Gene Details

Gene
ULBP1
Gene Name
UL16 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317089.1 659 Intron NP_001304018.1
NM_025218.3 659 Intron NP_079494.1
XM_017011322.1 659 Nonsense Mutation GAA,TAA E195* XP_016866811.1

View Full Product Details