Product Details
- SNP ID
-
rs77472767
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:59930295 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATCACAGAAATGCACAGGGCATA[A/C]GTTTTGTCTTAGAAGTCCCTACACT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LRRC9
PubMed Links
Gene Details
- Gene
- LRRC9
- Gene Name
- leucine rich repeat containing 9
There are no transcripts associated with this gene.
View Full Product Details