Product Details

SNP ID
rs77616515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149561582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGGGAGGAAATTCAAGTCATCA[A/G]TGTTCCATCTACTCATTGAAAACAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF37 PubMed Links

Gene Details

Gene
ARHGEF37
Gene Name
Rho guanine nucleotide exchange factor 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001669.2 Intron NP_001001669.2
XM_005268447.4 Intron XP_005268504.1
XM_005268448.1 Intron XP_005268505.1
XM_006714784.3 Intron XP_006714847.1
XM_011537642.2 Intron XP_011535944.1
XM_017009460.1 Intron XP_016864949.1
XM_017009461.1 Intron XP_016864950.1

View Full Product Details