Product Details

SNP ID
rs76403032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150784222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAGCCCTTACTGTGTACCAGAC[A/G]TTGTCCTAAGTGCTTTATGCGCATT
Phenotype
MIM: 608324
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMIM3 PubMed Links

Gene Details

Gene
SMIM3
Gene Name
small integral membrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032947.4 Intron NP_116565.3

View Full Product Details