Product Details

SNP ID
rs75681997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:148146968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGCAGTCACTGCTTGGGCTACCT[C/T]GCCATATGGCAAGTCAGGGTTTCTT
Phenotype
MIM: 605010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPINK14 PubMed Links

Gene Details

Gene
SPINK14
Gene Name
serine peptidase inhibitor, Kazal type 14 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001325.2 597 Intron NP_001001325.1
XM_017009466.1 597 Intron XP_016864955.1
XM_017009467.1 597 Silent Mutation CTC,CTT L66L XP_016864956.1
XM_017009468.1 597 UTR 5 XP_016864957.1
XM_017009469.1 597 Intron XP_016864958.1
Gene
SPINK5
Gene Name
serine peptidase inhibitor, Kazal type 5
There are no transcripts associated with this gene.

View Full Product Details