Product Details

SNP ID
rs79387527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:148385305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTAAATGGCTGTGTGCCCAGAG[G/T]TACACACTTTATATTTAGTCAAACA
Phenotype
MIM: 608533
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXO38 PubMed Links

Gene Details

Gene
FBXO38
Gene Name
F-box protein 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271723.1 Intron NP_001258652.1
NM_030793.4 Intron NP_110420.3
XM_005268513.1 Intron XP_005268570.1
XM_006714797.1 Intron XP_006714860.1
XM_011537684.2 Intron XP_011535986.1
XM_017009899.1 Intron XP_016865388.1
XM_017009900.1 Intron XP_016865389.1
XM_017009901.1 Intron XP_016865390.1
XM_017009902.1 Intron XP_016865391.1
Gene
LOC102546294
Gene Name
uncharacterized LOC102546294
There are no transcripts associated with this gene.

View Full Product Details