Product Details

SNP ID
rs76037300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55836121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCTTCCCCCTCTGACTCTCCTG[C/T]GCAGGGTCTGTTCCTGGTTTCCCAG
Phenotype
MIM: 606092 MIM: 601807
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC14 PubMed Links

Gene Details

Gene
DNAJC14
Gene Name
DnaJ heat shock protein family (Hsp40) member C14
There are no transcripts associated with this gene.

Gene
MMP19
Gene Name
matrix metallopeptidase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272101.1 2595 UTR 3 NP_001259030.1
NM_002429.5 2595 UTR 3 NP_002420.1
XM_006719401.3 2595 Intron XP_006719464.1
XM_011538359.1 2595 Intron XP_011536661.1
XM_017019308.1 2595 Intron XP_016874797.1
XM_017019309.1 2595 Intron XP_016874798.1
Gene
TMEM198B
Gene Name
transmembrane protein 198B (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details