Product Details

SNP ID
rs75016149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196296090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGCTAAAAAATCCAAGAATGAA[G/T]AATTATCAACAATCTAACAAGGACC
Phenotype
MIM: 123695
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCYT1A PubMed Links

Gene Details

Gene
PCYT1A
Gene Name
phosphate cytidylyltransferase 1, choline, alpha
There are no transcripts associated with this gene.

Gene
TCTEX1D2
Gene Name
Tctex1 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152773.4 Intron NP_689986.2

View Full Product Details