Product Details

SNP ID
rs75305282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10678243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTCTCATCCTTCTGAGCCTCTG[C/T]GGACACATCTAGAAAAATGGGGATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C8orf74 PubMed Links

Gene Details

Gene
C8orf74
Gene Name
chromosome 8 open reading frame 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040032.1 Intron NP_001035121.1
XM_011543816.2 Intron XP_011542118.1

View Full Product Details