Product Details

SNP ID
rs75969098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22689160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAAATCATTTAAAGCTGTATAC[A/G]CCTACAAATTCTCTTTATTCCGGTG
Phenotype
MIM: 602419
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EGR3 PubMed Links

Gene Details

Gene
EGR3
Gene Name
early growth response 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199880.1 2786 UTR 3 NP_001186809.1
NM_001199881.1 2786 UTR 3 NP_001186810.1
NM_004430.2 2786 UTR 3 NP_004421.2
XM_005273425.3 2786 UTR 3 XP_005273482.1
XM_005273426.3 2786 UTR 3 XP_005273483.1
XM_011544429.2 2786 UTR 3 XP_011542731.1

View Full Product Details