Product Details

SNP ID
rs74869265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22115482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAAAAGGGGCACAGGGTGGGGG[A/G]GATGCCAGCCTGAGAAGGACAGGTG
Phenotype
MIM: 602302 MIM: 615791
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HR PubMed Links

Gene Details

Gene
HR
Gene Name
hair growth associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005144.4 4453 UTR 3 NP_005135.2
NM_018411.4 4453 UTR 3 NP_060881.2
XM_005273569.1 4453 UTR 3 XP_005273626.1
XM_006716367.1 4453 UTR 3 XP_006716430.1
Gene
NUDT18
Gene Name
nudix hydrolase 18
There are no transcripts associated with this gene.

View Full Product Details