Product Details

SNP ID
rs112139659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:157543485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTCATTGCCTGAGGTTGGGCCC[G/A]AACACGGCGGTACATTTCCTTGAGG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
PQLC2L PubMed Links

Gene Details

Gene
PQLC2L
Gene Name
PQ loop repeat containing 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099777.3 142 UTR 5 NP_001093247.1
NM_001130001.2 142 UTR 5 NP_001123473.1
NM_001130002.2 142 UTR 5 NP_001123474.1
NM_001243729.1 142 UTR 5 NP_001230658.1
XM_017005754.1 142 UTR 5 XP_016861243.1

View Full Product Details