Product Details
- SNP ID
-
rs114667483
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:34657322 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGTCGGTTTAGTTTCTGCGGTTT[G/T]CAGGGTTGACATCCTAACTCCTTTG
- Phenotype
-
MIM: 606586
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105374753
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4024086] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105374753
- Gene Name
- uncharacterized LOC105374753
There are no transcripts associated with this gene.
- Gene
- RAI14
- Gene Name
- retinoic acid induced 14
View Full Product Details