Product Details

SNP ID
rs116778254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43121108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGAGGGACGGAGAGGAAGGAGG[G/A]CGACCCACGAGCTAGGTAAGGCGGG
Phenotype
MIM: 604073
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ZNF131 PubMed Links

Gene Details

Gene
ZNF131
Gene Name
zinc finger protein 131
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005248359.2 189 UTR 5 XP_005248416.1
XM_005248360.2 189 Intron XP_005248417.1
XM_005248361.2 189 Intron XP_005248418.1
XM_005248362.4 189 UTR 5 XP_005248419.1
XM_005248363.4 189 Intron XP_005248420.1
XM_005248365.3 189 UTR 5 XP_005248422.1
XM_005248367.2 189 UTR 5 XP_005248424.1
XM_011514123.2 189 Intron XP_011512425.1
XM_011514124.2 189 Intron XP_011512426.1
XM_017009830.1 189 Intron XP_016865319.1
XM_017009831.1 189 Intron XP_016865320.1
XM_017009832.1 189 Intron XP_016865321.1
XM_017009833.1 189 Intron XP_016865322.1
XM_017009834.1 189 UTR 5 XP_016865323.1
XM_017009835.1 189 Intron XP_016865324.1
XM_017009836.1 189 Intron XP_016865325.1
XM_017009837.1 189 Intron XP_016865326.1
XM_017009838.1 189 Intron XP_016865327.1
XM_017009839.1 189 Intron XP_016865328.1
XM_017009840.1 189 Intron XP_016865329.1
XM_017009841.1 189 UTR 5 XP_016865330.1
XM_017009842.1 189 Intron XP_016865331.1
XM_017009843.1 189 Intron XP_016865332.1

View Full Product Details