Product Details

SNP ID
rs115972924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:62313672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCCAGCCTCAGAGTTCTTAATA[C/T]AGCATTATCAGCCCTAATAGCCCTA
Phenotype
MIM: 602591
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF2A PubMed Links

Gene Details

Gene
KIF2A
Gene Name
kinesin family member 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098511.2 Intron NP_001091981.1
NM_001243952.1 Intron NP_001230881.1
NM_001243953.1 Intron NP_001230882.1
NM_004520.4 Intron NP_004511.2

View Full Product Details