Product Details

SNP ID
rs113081954
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:83054080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGTAAACTTGGAAAAGTTAATT[T/C]ATTCAATCTCTATTTTGTCCTAACT
Phenotype
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
TMEM167A PubMed Links

Gene Details

Gene
TMEM167A
Gene Name
transmembrane protein 167A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174909.4 Intron NP_777569.1

View Full Product Details