Product Details

SNP ID
rs117793191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35833063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTAAATAATTTGTTTATTGTAC[A/G]GCATTTACAAAGAAAACAGACAATG
Phenotype
MIM: 609427 MIM: 601939
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LHFPL5 PubMed Links

Gene Details

Gene
LHFPL5
Gene Name
lipoma HMGIC fusion partner-like 5
There are no transcripts associated with this gene.

Gene
SRPK1
Gene Name
SRSF protein kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003137.4 4333 UTR 3 NP_003128.3

View Full Product Details