Product Details

SNP ID
rs111627205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:32487272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGGAAACTCATTCACACTTCAG[A/G]TCCTTCTCCTCCAGGAACCAGCTGC
Phenotype
MIM: 612927 MIM: 607285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AVL9 PubMed Links

Gene Details

Gene
AVL9
Gene Name
AVL9 cell migration associated
There are no transcripts associated with this gene.

Gene
LSM5
Gene Name
LSM5 homolog, U6 small nuclear RNA and mRNA degradation associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130710.1 311 Missense Mutation CCT,TCT P60S NP_001124182.1
NM_001139499.1 311 Missense Mutation CCT,TCT P60S NP_001132971.1
NM_012322.2 311 Missense Mutation CCT,TCT P89S NP_036454.1

View Full Product Details