Product Details

SNP ID
rs114781832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149715251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTCTCTCCAGCTCTCCGCCGCC[G/T]GTACCCACGGCCCAGTCTCCACCTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRBA1 PubMed Links

Gene Details

Gene
KRBA1
Gene Name
KRAB-A domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290187.1 134 Intron NP_001277116.1
NM_032534.3 134 UTR 5 NP_115923.2
XM_011516636.2 134 Missense Mutation GGT,TGT G9C XP_011514938.1
XM_011516637.2 134 Missense Mutation GGT,TGT G9C XP_011514939.1
XM_011516638.2 134 Missense Mutation GGT,TGT G9C XP_011514940.1
XM_011516639.2 134 Missense Mutation GGT,TGT G9C XP_011514941.1
XM_011516640.2 134 Missense Mutation GGT,TGT G9C XP_011514942.1
XM_011516641.2 134 Intron XP_011514943.1
XM_011516642.2 134 UTR 5 XP_011514944.1
XM_011516643.2 134 Intron XP_011514945.1

View Full Product Details