Product Details
- SNP ID
-
rs111889356
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:95405465 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTAGTCACTGTAGGCTTCTCAGA[C/T]AGAATGTTCTGGATGCGGAGAACCT
- Phenotype
-
MIM: 602447
MIM: 602720
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PON2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7493] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PON2
- Gene Name
- paraoxonase 2
- Gene
- PON3
- Gene Name
- paraoxonase 3
There are no transcripts associated with this gene.
View Full Product Details