Product Details

SNP ID
rs117376828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102465915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGACCTGTCTAACAACCACCTGG[A/G]GACGCTGCCGGACAACCTGGGCCTG
Phenotype
MIM: 613302 MIM: 615167 MIM: 610929 MIM: 604150
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALKBH4 PubMed Links

Gene Details

Gene
ALKBH4
Gene Name
alkB homolog 4, lysine demethylase
There are no transcripts associated with this gene.

Gene
LRWD1
Gene Name
leucine rich repeats and WD repeat domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317721.1 395 UTR 5 NP_001304650.1
NM_152892.2 395 Missense Mutation GAG,GGG E60G NP_690852.1
XM_017011845.1 395 UTR 5 XP_016867334.1
Gene
MIR4467
Gene Name
microRNA 4467
There are no transcripts associated with this gene.

Gene
MIR5090
Gene Name
microRNA 5090
There are no transcripts associated with this gene.

Gene
ORAI2
Gene Name
ORAI calcium release-activated calcium modulator 2
There are no transcripts associated with this gene.

Gene
POLR2J
Gene Name
RNA polymerase II subunit J
There are no transcripts associated with this gene.

View Full Product Details