Product Details
- SNP ID
-
rs116121136
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:5231609 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGAAAGCCTAGCAGCAGAGCTGA[A/G]GGGATGTGGTCCCCGATTTGGAAAA
- Phenotype
-
MIM: 600910
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
INSL4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7852875] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- INSL4
- Gene Name
- insulin like 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002195.1 |
191 |
Missense Mutation |
AAG,AGG |
K29R |
NP_002186.1 |
View Full Product Details