Product Details

SNP ID
rs112468868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:35141073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCATTTACTGTATATTTTCACTG[C/T]GCCAGGCATTATGTTAAATATTTTC
Phenotype
MIM: 107269
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD44 PubMed Links

Gene Details

Gene
CD44
Gene Name
CD44 molecule (Indian blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000610.3 Intron NP_000601.3
NM_001001389.1 Intron NP_001001389.1
NM_001001390.1 Intron NP_001001390.1
NM_001001391.1 Intron NP_001001391.1
NM_001001392.1 Intron NP_001001392.1
NM_001202555.1 Intron NP_001189484.1
NM_001202556.1 Intron NP_001189485.1
NM_001202557.1 Intron NP_001189486.1
XM_005253231.2 Intron XP_005253288.1
XM_005253232.2 Intron XP_005253289.1
XM_005253233.2 Intron XP_005253290.1
XM_005253234.2 Intron XP_005253291.1
XM_005253235.2 Intron XP_005253292.1
XM_005253238.2 Intron XP_005253295.1
XM_005253239.2 Intron XP_005253296.1
XM_005253240.2 Intron XP_005253297.1
XM_006718388.1 Intron XP_006718451.1
XM_006718390.3 Intron XP_006718453.1
XM_011520482.1 Intron XP_011518784.1
XM_011520483.1 Intron XP_011518785.1
XM_011520484.1 Intron XP_011518786.1
XM_011520485.1 Intron XP_011518787.1
XM_011520486.1 Intron XP_011518788.1
XM_011520487.2 Intron XP_011518789.1
XM_011520488.1 Intron XP_011518790.1
XM_011520489.2 Intron XP_011518791.1
XM_017018583.1 Intron XP_016874072.1
XM_017018584.1 Intron XP_016874073.1
XM_017018585.1 Intron XP_016874074.1
Gene
LOC100507144
Gene Name
uncharacterized LOC100507144
There are no transcripts associated with this gene.

View Full Product Details