Product Details
- SNP ID
-
rs112306737
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:46615591 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCAGAAGCCAGCTATCTTTGTGC[A/G]TACCCGCTTCAAACTGACTACTGAG
- Phenotype
-
MIM: 615088
MIM: 615086
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATG13
PubMed Links
Gene Details
- Gene
- ATG13
- Gene Name
- autophagy related 13
There are no transcripts associated with this gene.
- Gene
- HARBI1
- Gene Name
- harbinger transposase derived 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173811.3 |
1167 |
Missense Mutation |
ACG,ATG |
T216M |
NP_776172.1 |
XM_011520025.1 |
1167 |
Missense Mutation |
ACG,ATG |
T216M |
XP_011518327.1 |
XM_011520026.2 |
1167 |
Missense Mutation |
ACG,ATG |
T216M |
XP_011518328.1 |
XM_011520027.1 |
1167 |
Missense Mutation |
ACG,ATG |
T216M |
XP_011518329.1 |
XM_011520028.2 |
1167 |
Missense Mutation |
ACG,ATG |
T216M |
XP_011518330.1 |
XM_011520029.2 |
1167 |
Missense Mutation |
ACG,ATG |
T216M |
XP_011518331.1 |
XM_017017602.1 |
1167 |
Missense Mutation |
ACG,ATG |
T216M |
XP_016873091.1 |
XM_017017603.1 |
1167 |
Missense Mutation |
ACG,ATG |
T216M |
XP_016873092.1 |
XM_017017604.1 |
1167 |
Missense Mutation |
ACG,ATG |
T216M |
XP_016873093.1 |
XM_017017605.1 |
1167 |
Missense Mutation |
ACG,ATG |
T216M |
XP_016873094.1 |
XM_017017606.1 |
1167 |
Missense Mutation |
ACG,ATG |
T216M |
XP_016873095.1 |
View Full Product Details