Product Details
- SNP ID
-
rs121434363
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:47410315 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACACCTGGATCTGCTCCCTCCTGG[A/G]TTCCCTCATGGTGGGGCTCAGTGGG
- Phenotype
-
MIM: 186852
MIM: 608735
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4487
PubMed Links
Gene Details
- Gene
- MIR4487
- Gene Name
- microRNA 4487
There are no transcripts associated with this gene.
- Gene
- PSMC3
- Gene Name
- proteasome 26S subunit, ATPase 3
There are no transcripts associated with this gene.
- Gene
- SLC39A13
- Gene Name
- solute carrier family 39 member 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001128225.2 |
276 |
Missense Mutation |
GAT,GGT |
D74G |
NP_001121697.1 |
NM_152264.4 |
276 |
Missense Mutation |
GAT,GGT |
D74G |
NP_689477.2 |
XM_006718381.3 |
276 |
Missense Mutation |
GAT,GGT |
D89G |
XP_006718444.1 |
XM_006718383.3 |
276 |
Missense Mutation |
GAT,GGT |
D89G |
XP_006718446.1 |
XM_006718384.2 |
276 |
Missense Mutation |
GAT,GGT |
D89G |
XP_006718447.1 |
XM_006718385.2 |
276 |
Missense Mutation |
GAT,GGT |
D89G |
XP_006718448.1 |
XM_011520466.1 |
276 |
Missense Mutation |
GAT,GGT |
D89G |
XP_011518768.1 |
XM_011520467.1 |
276 |
Missense Mutation |
GAT,GGT |
D74G |
XP_011518769.1 |
XM_011520468.2 |
276 |
Missense Mutation |
GAT,GGT |
D74G |
XP_011518770.1 |
XM_011520469.1 |
276 |
Missense Mutation |
GAT,GGT |
D89G |
XP_011518771.1 |
XM_011520470.1 |
276 |
Missense Mutation |
GAT,GGT |
D74G |
XP_011518772.1 |
XM_017018540.1 |
276 |
Missense Mutation |
GAT,GGT |
D74G |
XP_016874029.1 |
XM_017018541.1 |
276 |
Missense Mutation |
GAT,GGT |
D74G |
XP_016874030.1 |
XM_017018542.1 |
276 |
Missense Mutation |
GAT,GGT |
D74G |
XP_016874031.1 |
View Full Product Details