Product Details

SNP ID
rs116778909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6002430 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGATGTCATCACAAGAGAGTTTG[A/G]ACACAGACAGGTTACTGCAGATGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR56A4 PubMed Links

Gene Details

Gene
OR56A4
Gene Name
olfactory receptor family 56 subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005179.2 719 Missense Mutation TCC,TTC S240F NP_001005179.2

View Full Product Details