Product Details
- SNP ID
-
rs117241472
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:7486046 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCTTGGATAAGTAACAGGCTTCC[C/T]AGAAGTACCTTATTTTTACCCATGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OLFML1
PubMed Links
Gene Details
- Gene
- OLFML1
- Gene Name
- olfactomedin like 1
View Full Product Details