Product Details

SNP ID
rs111256472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1938963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCCAAGAAACGAAAAGGTCTTG[T/C]TCAGAGCCGGGGCCTTGGGAGTGTC
Phenotype
MIM: 606216
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
MSRB1 PubMed Links

Gene Details

Gene
MSRB1
Gene Name
methionine sulfoxide reductase B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016332.2 637 UTR 3 NP_057416.1
Gene
RPL3L
Gene Name
ribosomal protein L3 like
There are no transcripts associated with this gene.

View Full Product Details