Product Details

SNP ID
rs117014569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87409846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAGGAACAGAACAAAACAAAGT[C/T]GACATTTTCCCTAGCCCAGGGGTGT
Phenotype
MIM: 609604
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP1LC3B PubMed Links

Gene Details

Gene
MAP1LC3B
Gene Name
microtubule associated protein 1 light chain 3 beta
There are no transcripts associated with this gene.

Gene
ZCCHC14
Gene Name
zinc finger CCHC-type containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015144.2 3311 UTR 3 NP_055959.1
XM_005255858.3 3311 Intron XP_005255915.2
XM_011522964.2 3311 Intron XP_011521266.1
XM_017023082.1 3311 Intron XP_016878571.1

View Full Product Details