Product Details

SNP ID
rs117157555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2757524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCGCCCGACCACAGCGCCTGGCT[C/T]AAGTATGGGTGTGTGACCTAGGTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC92B PubMed Links

Gene Details

Gene
CCDC92B
Gene Name
coiled-coil domain containing 92B
There are no transcripts associated with this gene.

Gene
MIR1253
Gene Name
microRNA 1253
There are no transcripts associated with this gene.

Gene
RAP1GAP2
Gene Name
RAP1 GTPase activating protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100398.1 Intron NP_001093868.1
NM_015085.4 Intron NP_055900.4
XM_005256542.2 Intron XP_005256599.1
XM_006721477.3 Intron XP_006721540.1
XM_011523738.1 Intron XP_011522040.1
XM_011523739.1 Intron XP_011522041.1
XM_011523740.2 Intron XP_011522042.1
XM_011523741.2 Intron XP_011522043.1
XM_011523742.2 Intron XP_011522044.1
XM_011523743.2 Intron XP_011522045.1
XM_011523744.2 Intron XP_011522046.1
XM_011523745.2 Intron XP_011522047.1
XM_017024370.1 Intron XP_016879859.1
XM_017024371.1 Intron XP_016879860.1

View Full Product Details