Product Details

SNP ID
rs117033540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58088999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAAAACCTGAGCTTTTGAGGTTC[G/T]GGGAGCTGAGGCTGAGGGATGGGGT
Phenotype
MIM: 608942
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DYNLL2 PubMed Links

Gene Details

Gene
DYNLL2
Gene Name
dynein light chain LC8-type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080677.2 Intron NP_542408.1
Gene
LOC101927666
Gene Name
uncharacterized LOC101927666
There are no transcripts associated with this gene.

View Full Product Details