Product Details

SNP ID
rs116245925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63703427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGTCTCAAAAGCCTTGGAGCAG[G/T]GAAGTGTCTCTCCAGGATTTTCTTT
Phenotype
MIM: 602539 MIM: 608626
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LIMD2 PubMed Links

Gene Details

Gene
LIMD2
Gene Name
LIM domain containing 2
There are no transcripts associated with this gene.

Gene
LOC729683
Gene Name
uncharacterized LOC729683
There are no transcripts associated with this gene.

Gene
MAP3K3
Gene Name
mitogen-activated protein kinase kinase kinase 3
There are no transcripts associated with this gene.

Gene
STRADA
Gene Name
STE20-related kinase adaptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003786.2 1564 UTR 3 NP_001003786.1
NM_001003787.2 1564 UTR 3 NP_001003787.1
NM_001003788.2 1564 UTR 3 NP_001003788.1
NM_001165969.1 1564 UTR 3 NP_001159441.1
NM_001165970.1 1564 UTR 3 NP_001159442.1
NM_153335.5 1564 UTR 3 NP_699166.2
XM_005257797.2 1564 UTR 3 XP_005257854.1
XM_005257798.2 1564 UTR 3 XP_005257855.1
XM_005257799.2 1564 UTR 3 XP_005257856.1
XM_005257800.2 1564 UTR 3 XP_005257857.1
XM_005257801.4 1564 Intron XP_005257858.1
XM_005257803.4 1564 Intron XP_005257860.1
XM_011525466.2 1564 UTR 3 XP_011523768.1
XM_011525467.2 1564 UTR 3 XP_011523769.1
XM_017025312.1 1564 UTR 3 XP_016880801.1
XM_017025313.1 1564 UTR 3 XP_016880802.1
XM_017025314.1 1564 Intron XP_016880803.1

View Full Product Details