Product Details
- SNP ID
-
rs114671607
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41139873 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGATAGTTTCAGGCAAGGAGCTCA[A/G]CAGCAAGAGGAGGCACAGCACAAGG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRTAP4-5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs551614044] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRTAP4-5
- Gene Name
- keratin associated protein 4-5
There are no transcripts associated with this gene.
- Gene
- KRTAP4-6
- Gene Name
- keratin associated protein 4-6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030976.1 |
615 |
Silent Mutation |
TGC,TGT |
C205C |
NP_112238.1 |
View Full Product Details